1-Phenethylimidazole 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 1-(Phenylethyl)imidazole, 1-Phenethyl-1H-imidazole

A557194-250MG A557194-25G A557194-5G
76830-598EA 7 USD
76830-598 76830-600 77684-404
1-Phenethylimidazole 98%, Ambeed.Inc
1-Phenethylimidazole

Precaución: For research use only.

Formula: C₁₁H₁₂N₂
MDL Number: MFCD11977663
CAS Number: 49823-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR