4-Formylphenyl-β-D-allopyranoside 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 4-(((2S,3R,4R,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)benzaldehyde, 4-(β-D-Allopyranosyloxy)benzaldehyde, Helicide

A556507-10G A556507-25G A556507-1G
76830-594EA 81.69 USD
76830-594 76830-596 77684-402
4-Formylphenyl-β-D-allopyranoside 97%, Ambeed.Inc
4-Formylphenyl-beta-D-allopyranoside

Precaución: For research use only.

Formula: C₁₃H₁₆O₇
MW: 284.26 g/mol
MDL Number: MFCD00210992
CAS Number: 80154-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR