VER-155008 98%

Supplier: Ambeed
Warning

Synonyms: 5'-O-[(4-Cyanophenyl)methyl]-8-[[(3,4-dichlorophenyl)methyl]amino]-adenosine, 4-((((2R,3S,4R,5R)-5-(6-Amino-8-((3,4-dichlorobenzyl)amino)-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)methyl)benzonitrile

A556276-10MG A556276-1MG A556276-50MG A556276-5MG
76830-580EA 53.05 USD
76830-580 76830-582 76830-584 76830-586
VER-155008 98%
VER-155008

Precaución: for Research Use Only

Formula: C₂₅H₂₃Cl₂N₇O₄
MW: 556.40 g/mol
MDL Number: MFCD18086892
CAS Number: 1134156-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR