7-Chloroimidazo[1,2-a]pyrimidin-5(1H)-one 97%

Supplier: Ambeed
Warning

A554083-100MG A554083-1G A554083-250MG
76830-570EA 41 USD
76830-570 77551-668 77886-368
7-Chloroimidazo[1,2-a]pyrimidin-5(1H)-one 97%
7-Chloroimidazo[1,2-a]pyrimidin-5(1H)-one

Precaución: for Research Use Only

Formula: C₆H₄ClN₃O
MDL Number: MFCD14706869
CAS Number: 57473-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR