4-(4-Chlorophenyl)piperidine-4-carbonitrile 98%

Supplier: Ambeed
Warning

A553777-100MG A553777-1G A553777-250MG
76830-564EA 87 USD
76830-564 76830-566 76830-568
4-(4-Chlorophenyl)piperidine-4-carbonitrile 98%
4-(4-Chlorophenyl)piperidine-4-carbonitrile

Precaución: for Research Use Only

Formula: C₁₂H₁₃ClN₂
MW: 220.70 g/mol
MDL Number: MFCD11878513
CAS Number: 91721-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR