2-(Butylamino)-5-pyrimidineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: N-Butyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

A553198-1G A553198-250MG A553198-5G
76830-558EA 135.8 USD
76830-558 76830-560 76830-562
2-(Butylamino)-5-pyrimidineboronic acid pinacol ester 98%
2-(Butylamino)-5-pyrimidineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₄BN₃O₂
MW: 277.1700 g/mol
MDL Number: MFCD12546558
CAS Number: 1218791-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR