4-tert-Butylpyrocatechol 98%

Supplier: Ambeed
Danger

Synonyms: 4-(tert-Butyl)benzene-1,2-diol, 4-tert-Butylcatechol

A552712-25G A552712-500G A552712-1000G
76830-554EA 6.37 USD
76830-554 76830-556 77551-664
4-tert-Butylpyrocatechol 98%
4-tert-Butylpyrocatechol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
MDL Number: MFCD00002201
CAS Number: 98-29-3
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR