5-(Ethoxycarbonyl)thiophene-3-boronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (5-(Ethoxycarbonyl)thiophen-3-yl)boronic acid, Ethyl 4-boronothiophene-2-carboxylate, 4-Borono-2-(ethoxycarbonyl)thiophene

A552032-100MG A552032-1G A552032-250MG
76830-544EA 37 USD
76830-544 76830-546 76830-548
5-(Ethoxycarbonyl)thiophene-3-boronic acid 96%
5-(Ethoxycarbonyl)thiophene-3-boronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₄S
MDL Number: MFCD09800891
CAS Number: 957121-19-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR