1-(4-Bromophenyl)adamantane 95%

Supplier: Ambeed
Warning

A549971-1G A549971-25G A549971-5G
76830-542EA 20 USD
76830-542 77551-662 77886-365
1-(4-Bromophenyl)adamantane 95%
1-(4-Bromophenyl)adamantane

Precaución: for Research Use Only

Formula: C₁₆H₁₉Br
MDL Number: MFCD01882238
CAS Number: 2245-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR