2-Chloro-4-nitro-1-(trifluoromethoxy)benzene 98%

Supplier: Ambeed
Warning

A548611-10G A548611-25G
76830-536EA 6 USD
76830-536 76830-538 76830-540
2-Chloro-4-nitro-1-(trifluoromethoxy)benzene 98%
2-Chloro-4-nitro-1-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃ClF₃NO₃
MDL Number: MFCD12032157
CAS Number: 158579-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR