5,10,15,20-Tetrakis(4-aminophenyl)porphyrin 98%

Supplier: Ambeed
Warning

Synonyms: 5,10,15,20-Tetrakis(4-aminophenyl)-21H,23H-porphine

A545277-100MG A545277-1G A545277-250MG A545277-25G
76830-530EA 15 USD
76830-530 76830-532 76830-534 77886-361
5,10,15,20-Tetrakis(4-aminophenyl)porphyrin 98%
5,10,15,20-Tetrakis(4-aminophenyl)porphyrin

Precaución: for Research Use Only

Formula: C₄₄H₃₄N₈
MW: 674.81 g/mol
MDL Number: MFCD00191497
CAS Number: 22112-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR