5-Fluoroquinazoline-2,4(1H,3H)-dione 95%

Supplier: Ambeed
Warning

A543617-100MG A543617-250MG
76830-526EA 23.34 USD
76830-526 76830-528
5-Fluoroquinazoline-2,4(1H,3H)-dione 95%
5-Fluoroquinazoline-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₈H₅FN₂O₂
MW: 180.136 g/mol
MDL Number: MFCD09954841
CAS Number: 192570-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR