N-TIPS pyrrole-3-boronic acid pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(4,4,5,5-TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)-1-[TRIS(1-METHYLETHYL)SILYL]-1H-PYRROLE, 3-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1-[tris(propan-2-yl)silyl]-1H-pyrrole, 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(triisopropylsilyl)-1H-pyrrole
A541421-1G
A541421-250MG
A541421-5G
76830-508EA
20
USD
76830-508
76830-510
76830-512
N-TIPS pyrrole-3-boronic acid pinacol ester 98%
N-TIPS pyrrole-3-boronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₉H₃₆BNO₂Si |
MDL Number:
MFCD08063117 CAS Number: 365564-11-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...