2-Methyl-2H-indazole-6-boronic acid 95+%

Supplier: Ambeed
Warning

Synonyms: (2-Methyl-2H-indazol-6-yl)boronic acid

A540662-100MG A540662-1G A540662-250MG
76830-498EA 70.02 USD
76830-498 76830-500 76830-502
2-Methyl-2H-indazole-6-boronic acid 95+%
2-Methyl-2H-indazole-6-boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₂
MW: 175.9800 g/mol
MDL Number: MFCD09870057
CAS Number: 1001907-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR