3,6-Dimethylimidazo[2,1-b]thiazole 95%

Supplier: Ambeed
Warning

A535715-100MG A535715-1G A535715-250MG
76830-446EA 26 USD
76830-446 76830-448 76830-450
3,6-Dimethylimidazo[2,1-b]thiazole 95%
3,6-Dimethylimidazo[2,1-b]thiazole

Precaución: for Research Use Only

Formula: C₇H₈N₂S
MDL Number: MFCD03556234
CAS Number: 25944-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR