1-(3-Fluorophenyl)cyclopropanecarboxylic acid 95%

Supplier: Ambeed
Warning

A534761-100MG A534761-250MG
76830-436EA 35.01 USD
76830-436 76830-438
1-(3-Fluorophenyl)cyclopropanecarboxylic acid 95%
1-(3-Fluorophenyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉FO₂
MW: 180.18 g/mol
MDL Number: MFCD07374443
CAS Number: 248588-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR