Methyl 1-acetyl-1H-indazole-5-carboxylate 95%

Supplier: Ambeed
Warning

A532663-250MG A532663-5G
76830-422EA 20.16 USD
76830-422 76830-424
Methyl 1-acetyl-1H-indazole-5-carboxylate 95%
Methyl 1-acetyl-1H-indazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O₃
MW: 218.21 g/mol
MDL Number: MFCD09838842
CAS Number: 239075-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR