4,6-Dichloro-1H-indazole 97%

Supplier: Ambeed
Warning

A531989-100MG A531989-250MG
76830-418EA 43.5 USD
76830-418 76830-420
4,6-Dichloro-1H-indazole 97%
4,6-Dichloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂
MW: 187.026 g/mol
MDL Number: MFCD07781384
CAS Number: 885519-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR