1-(Phenylsulphonyl)-5-indoleboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (1-(Phenylsulphonyl)-1H-indol-5-yl)boronic acid

A531714-1G A531714-250MG A531714-5G
76830-412EA 100 USD
76830-412 76830-414 76830-416
1-(Phenylsulphonyl)-5-indoleboronic acid 95%
1-(Phenylsulphonyl)-5-indoleboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂BNO₄S
MW: 301.1300 g/mol
MDL Number: MFCD03840606
CAS Number: 480438-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR