Ethyl 3-bromo-2,2-difluoropropionate 98%

Supplier: Ambeed
Danger

Synonyms: Ethyl 3-bromo-2,2-difluoropropanoate

A529471-1G A529471-5G
76830-408EA 55 USD
76830-408 77886-333
Ethyl 3-bromo-2,2-difluoropropionate 98%
Ethyl 3-bromo-2,2-difluoropropionate

Precaución: for Research Use Only

Formula: C₅H₇BrF₂O₂
MDL Number: MFCD15144891
CAS Number: 111773-24-1
UN: 3272
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR