8-Chloroquinazolin-4(3H)-one 95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 8-Chloroquinazolin-4-ol

A528935-250MG A528935-5G A528935-1G
76830-404EA 41.38 USD
76830-404 76830-406 77684-368
8-Chloroquinazolin-4(3H)-one 95%, Ambeed.Inc
8-Chloroquinazolin-4(3H)-one

Precaución: For research use only.

Formula: C₈H₅ClN₂O
MW: 180.591 g/mol
MDL Number: MFCD09954861
CAS Number: 101494-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR