6-Acetyl-1-methyl-1H-indazole 95%

Supplier: Ambeed
Warning

Synonyms: 1-(1-Methyl-1H-indazol-6-yl)ethan-1-one, 1-(1-Methyl-1H-indazol-6-yl)ethanone, 6-Ethanoyl-1-methyl-1H-indazole

A526782-1G A526782-250MG
76830-394EA 315.09 USD
76830-394 76830-396
6-Acetyl-1-methyl-1H-indazole 95%
6-Acetyl-1-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O
MW: 174.199 g/mol
MDL Number: MFCD11869768
CAS Number: 1159511-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR