4-[(Isobutylcarbamoyl)amino]benzeneboronic acid, pinacol ester 98%

Supplier: Ambeed
Danger

Synonyms: 1-Isobutyl-3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea, 4-(3-Isobutylureido)phenylboronic acid, pinacol ester

A525488-1G A525488-250MG A525488-5G
76830-388EA 128 USD
76830-388 76830-390 76830-392
4-[(Isobutylcarbamoyl)amino]benzeneboronic acid, pinacol ester 98%
4-[(Isobutylcarbamoyl)amino]benzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₇BN₂O₃
MDL Number: MFCD09027289
CAS Number: 874291-03-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR