1-Aminocyclopropanecarboxylic acid hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 1-azanylcyclopropane-1-carboxylic acid hydrochloride, ACPC, 1-aminocyclopropane-1-carboxylic acid hydrochloride, Cyclopropanecarboxylic acid, 1-amino-, hydrochloride

A521191-100G A521191-25G A521191-5G
76830-372EA 246.13 USD
76830-372 76830-374 76830-376
1-Aminocyclopropanecarboxylic acid hydrochloride 98%
1-Aminocyclopropanecarboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₄H₈ClNO₂
MW: 137.57 g/mol
MDL Number: MFCD00012545
CAS Number: 68781-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR