5-Chloro-2-isopropoxy-3-pyridineboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (5-Chloro-2-isopropoxypyridin-3-yl)boronic acid, 5-Chloro-2-isopropoxypyridine-3-boronic acid

A521051-100MG A521051-1G A521051-250MG A521051-5G
76830-364EA 32 USD
76830-364 76830-366 76830-368 76830-370
5-Chloro-2-isopropoxy-3-pyridineboronic acid 96%
5-Chloro-2-isopropoxy-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BClNO₃
MDL Number: MFCD13195662
CAS Number: 1217501-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR