7-Boc-2-hydroxy-7-azaspiro[3.5]nonane 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-hydroxy-7-azaspiro[3.5]nonane-7-carboxylate

A520953-1G A520953-250MG A520953-25G A520953-5G A520953-100G
76830-356EA 40.32 USD
76830-356 76830-358 76830-360 76830-362 77684-351
7-Boc-2-hydroxy-7-azaspiro[3.5]nonane 98%
7-Boc-2-hydroxy-7-azaspiro[3.5]nonane

Precaución: For research use only.

Formula: C₁₃H₂₃NO₃
MW: 241.327 g/mol
MDL Number: MFCD14584466
CAS Number: 240401-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR