1-(3-Methoxyphenyl)-1H-pyrazole-4-carbaldehyde 95%

Supplier: Ambeed
Warning

A518119-100MG A518119-1G A518119-250MG
76830-344EA 65.78 USD
76830-344 76830-346 76830-348
1-(3-Methoxyphenyl)-1H-pyrazole-4-carbaldehyde 95%
1-(3-Methoxyphenyl)-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O₂
MW: 202.20 g/mol
MDL Number: MFCD13874001
CAS Number: 1013835-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR