Ethyl 2-(2,4-difluorophenyl)-2,2-difluoroacetate 97%

Supplier: Ambeed
Warning

A516373-1G A516373-250MG
76830-334EA 143.23 USD
76830-334 76830-336
Ethyl 2-(2,4-difluorophenyl)-2,2-difluoroacetate 97%
Ethyl 2-(2,4-difluorophenyl)-2,2-difluoroacetate

Precaución: for Research Use Only

Formula: C₁₀H₈F₄O₂
MW: 236.16 g/mol
MDL Number: MFCD16618951
CAS Number: 1228957-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR