6-Chloro-1H-indole-2-carbaldehyde 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A516021-1G A516021-250MG A516021-100MG
76830-330EA 92.3 USD
76830-330 76830-332 77684-342
6-Chloro-1H-indole-2-carbaldehyde 97%, Ambeed.Inc
6-Chloro-1H-indole-2-carbaldehyde

Precaución: For research use only.

Formula: C₉H₆ClNO
MW: 179.603 g/mol
MDL Number: MFCD11040767
CAS Number: 53590-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR