(4-(2-Hydroxypropan-2-yl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A515753-100MG A515753-250MG A515753-5G
76830-326EA 7.43 USD
76830-326 76830-328 77613-624
(4-(2-Hydroxypropan-2-yl)phenyl)boronic acid 97%
(4-(2-Hydroxypropan-2-yl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₃
MW: 180.009 g/mol
MDL Number: MFCD09992905
CAS Number: 886593-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR