(R)-2-Amino-3,3-diphenylpropan-1-ol 95%

Supplier: Ambeed
Warning

A515232-100MG A515232-250MG
76830-322EA 95.49 USD
76830-322 76830-324
(R)-2-Amino-3,3-diphenylpropan-1-ol 95%
(R)-2-Amino-3,3-diphenylpropan-1-ol

Precaución: for Research Use Only

Formula: C₁₅H₁₇NO
MW: 227.30 g/mol
MDL Number: MFCD08063386
CAS Number: 171037-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR