2-Formylfuran-4-boronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carbaldehyde

A515018-100MG A515018-1G A515018-250MG A515018-5G
76830-316EA 7 USD
76830-316 76830-318 76830-320 77572-642
2-Formylfuran-4-boronic acid pinacol ester 97%
2-Formylfuran-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₅BO₄
MDL Number: MFCD06657890
CAS Number: 846023-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR