6-(Trifluoromethyl)pyridine-2-carboxamide 95%

Supplier: Ambeed
Warning

Synonyms: 6-(Trifluoromethyl)picolinamide

A513445-100G A513445-1G A513445-25G A513445-500G A513445-5G
76830-302EA 341.61 USD
76830-302 76830-304 76830-306 76830-308 76830-310
6-(Trifluoromethyl)pyridine-2-carboxamide 95%
6-(Trifluoromethyl)pyridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₇H₅F₃N₂O
MW: 190.123 g/mol
MDL Number: MFCD02180820
CAS Number: 22245-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR