4-Chloro-2,6-dimethylphenylboronic acid 98%

Supplier: Ambeed
Warning

A513354-1G A513354-250MG
76830-298EA 288.57 USD
76830-298 76830-300
4-Chloro-2,6-dimethylphenylboronic acid 98%
4-Chloro-2,6-dimethylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BClO₂
MW: 184.4278 g/mol
MDL Number: MFCD10566594
CAS Number: 1027045-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR