1,3-Dihydrobenzo[c]thiophene 95%

Supplier: Ambeed
Danger

A512780-100MG A512780-1G A512780-250MG
76830-292EA 122 USD
76830-292 76830-294 76830-296
1,3-Dihydrobenzo[c]thiophene 95%
1,3-Dihydrobenzo[c]thiophene

Precaución: for Research Use Only

Formula: C₈H₈S
MDL Number: MFCD18382467
CAS Number: 2471-92-3
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR