(2,4-Bis(methoxycarbonyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A510803-100MG A510803-1G A510803-250MG
76830-280EA 36 USD
76830-280 76830-282 76830-284
(2,4-Bis(methoxycarbonyl)phenyl)boronic acid 98%
(2,4-Bis(methoxycarbonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BO₆
MW: 238.00 g/mol
MDL Number: MFCD17015764
CAS Number: 1256354-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR