Tris(2-benzimidazolylmethyl)amine 95%

Supplier: Ambeed
Warning

A509121-100MG A509121-250MG A509121-5G
76830-274EA 26 USD
76830-274 76830-276 77886-303
Tris(2-benzimidazolylmethyl)amine 95%
Tris(2-benzimidazolylmethyl)amine

Precaución: for Research Use Only

Formula: C₂₄H₂₁N₇
MDL Number: MFCD00075516
CAS Number: 64019-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR