Methyl 7-fluoro-1H-indazole-4-carboxylate 95%

Supplier: Ambeed
Warning

A508994-100MG A508994-1G A508994-250MG
76830-268EA 72.15 USD
76830-268 76830-270 76830-272
Methyl 7-fluoro-1H-indazole-4-carboxylate 95%
Methyl 7-fluoro-1H-indazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇FN₂O₂
MW: 194.162 g/mol
MDL Number: MFCD21607513
CAS Number: 1079993-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR