1-(3-Bromophenyl)cyclopropanecarboxylic acid 96%

Supplier: Ambeed
Warning

A506643-100MG A506643-1G A506643-250MG A506643-25G A506643-5G
76830-252EA 8.49 USD
76830-252 76830-254 76830-256 76830-258 76830-260
1-(3-Bromophenyl)cyclopropanecarboxylic acid 96%
1-(3-Bromophenyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₂
MW: 241.0813 g/mol
MDL Number: MFCD07374441
CAS Number: 124276-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR