5-Bromo-2,4-dichloropyrimidine 98%

Supplier: Ambeed
Danger

A506344-10G A506344-1G A506344-25G A506344-5G
76830-244EA 8.49 USD
76830-244 76830-246 76830-248 76830-250
5-Bromo-2,4-dichloropyrimidine 98%
5-Bromo-2,4-dichloropyrimidine

Precaución: for Research Use Only

Formula: C₄HBrCl₂N₂
MW: 227.87 g/mol
MDL Number: MFCD00127818
CAS Number: 36082-50-5
UN: 2923
ADR: 8,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR