3-(2-Chloroethyl)oxazolidin-2-one 95%

Supplier: Ambeed
Danger

A504561-100MG A504561-1G A504561-250MG A504561-5G
76830-222EA 78.51 USD
76830-222 76830-224 76830-226 76830-228
3-(2-Chloroethyl)oxazolidin-2-one 95%
3-(2-Chloroethyl)oxazolidin-2-one

Precaución: for Research Use Only

Formula: C₅H₈ClNO₂
MW: 149.57 g/mol
MDL Number: MFCD00046094
CAS Number: 2508-01-2
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR