5,7-Difluorobenzo[d]thiazol-2-amine 97%

Supplier: Ambeed
Warning

A504281-100MG A504281-1G A504281-250MG
76830-216EA 19 USD
76830-216 76830-218 76830-220
5,7-Difluorobenzo[d]thiazol-2-amine 97%
5,7-Difluorobenzo[d]thiazol-2-amine

Precaución: for Research Use Only

Formula: C₇H₄F₂N₂S
MDL Number: MFCD08444544
CAS Number: 788124-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR