Cyclohexane-1,2,3,4,5,6-hexaol 98%

Supplier: Ambeed
Warning

A499991-1000G A499991-100G A499991-10G A499991-25G A499991-500G
76830-176EA 309 USD
76830-176 76830-178 76830-180 76830-182 76830-184
Cyclohexane-1,2,3,4,5,6-hexaol 98%
Cyclohexane-1,2,3,4,5,6-hexaol

Precaución: for Research Use Only

Formula: C₆H₁₂O₆
MDL Number: MFCD00077932
CAS Number: 6917-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR