(R)-3,3'-Bis(triphenylsilyl)-[1,1'-binaphthalene]-2,2'-diol ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
A498626-100MG
A498626-1G
A498626-250MG
A498626-50MG
76830-152EA
78.51
USD
76830-152
76830-154
76830-156
76830-158
(R)-3,3'-Bis(triphenylsilyl)-[1,1'-binaphthalene]-2,2'-diol ≥98%, ee 99%
(R)-3,3'-Bis(triphenylsilyl)-[1,1'-binaphthalene]-2,2'-diol
Precaución: for Research Use Only
Formula:
C₅₆H₄₂O₂Si₂ MW: 803.10 g/mol |
MDL Number:
MFCD09265078 CAS Number: 111822-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...