N2,N2-Dimethylpyrimidine-2,5-diamine 95%

Supplier: Ambeed
Warning

A497416-250MG A497416-5G A497416-1G
76830-148EA 96.55 USD
76830-148 76830-150 76878-026
N2,N2-Dimethylpyrimidine-2,5-diamine 95%
N2,N2-Dimethylpyrimidine-2,5-diamine

Precaución: for Research Use Only

Formula: C₆H₁₀N₄
MW: 138.17 g/mol
MDL Number: MFCD11656641
CAS Number: 56621-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR