2-(1-Trityl-1H-imidazol-4-yl)ethanamine 97%

Supplier: Ambeed
Danger

A497044-1G A497044-5G A497044-250MG
76830-138EA 38 USD
76830-138 77551-508 77886-286
2-(1-Trityl-1H-imidazol-4-yl)ethanamine 97%
2-(1-Trityl-1H-imidazol-4-yl)ethanamine

Precaución: for Research Use Only

Formula: C₂₄H₂₃N₃
MDL Number: MFCD06738986
CAS Number: 195053-92-0
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR