4-Bromo-2-(trifluoromethoxy)phenol 98%
Supplier: Ambeed
|
Warning
|
A496885-1G
A496885-250MG
A496885-25G
A496885-5G
76830-130EA
30.77
USD
76830-130
76830-132
76830-134
76830-136
4-Bromo-2-(trifluoromethoxy)phenol 98%
4-Bromo-2-(trifluoromethoxy)phenol
Precaución: for Research Use Only
Formula:
C₇H₄BrF₃O₂ |
MDL Number:
MFCD09037798 CAS Number: 690264-39-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...