4-Bromo-4'-butyl-1,1'-biphenyl 98%

Supplier: Ambeed
Warning

A496466-1G A496466-250MG A496466-5G
76830-124EA 7.43 USD
76830-124 76830-126 76830-128
4-Bromo-4'-butyl-1,1'-biphenyl 98%
4-Bromo-4'-butyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₆H₁₇Br
MW: 289.21 g/mol
MDL Number: MFCD11977251
CAS Number: 63619-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR