3-Propylbenzene-1,2-diol 95%

Supplier: Ambeed
Warning

A493658-1G A493658-250MG
76830-102EA 483.78 USD
76830-102 76830-104
3-Propylbenzene-1,2-diol 95%
3-Propylbenzene-1,2-diol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
MDL Number: MFCD00873219
CAS Number: 2896-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR