1,5-Dichloroisoquinoline 97%
Supplier: Ambeed
|
Warning
|
A492601-100MG
A492601-1G
A492601-250MG
76830-086EA
15
USD
76830-086
76830-088
76830-090
1,5-Dichloroisoquinoline 97%
1,5-Dichloroisoquinoline
Precaución: for Research Use Only
Formula:
C₉H₅Cl₂N |
MDL Number:
MFCD11846292 CAS Number: 70810-23-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...